Skip to Content
Merck
Boc-Thr-OH Novabiochem®

Boc-Thr-OH

Boc-Thr-OH Novabiochem®
Synonym(s):
Boc-Thr-OH
Empirical Formula (Hill Notation):
C9H17NO5
CAS Number:
Molecular Weight:
219.23

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Functional Group
Application
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing