Skip to Content
Merck
Boc-Ser-OMe 95%

Boc-Ser-OMe

Boc-Ser-OMe 95%
Synonym(s):
N-(tert-Butoxycarbonyl)-L-serine methyl ester, Boc-L-serine methyl ester
Linear Formula:
HOCH2CH[NHCO2C(CH3)3]CO2CH3
CAS Number:
Molecular Weight:
219.23

Products

Brand
Formula Weight
Physical Form
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing