Skip to Content
Merck
BIOshell A400 Protein C18 Column L × I.D. 3 cm × 4.6 mm, 3.4 μm particle size

BIOshell A400 Protein C18 (3.4 μm) HPLC Columns

BIOshell A400 Protein C18 Column L × I.D. 3 cm × 4.6 mm, 3.4 μm particle size

Products

Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Analyte Suitability
Separation Technique
ID (mm)
Length (cm)
Quality Segment
Package Size
Particle Size (µm)
Pore Size (Å)
Pore size (µm)
Technique
Showing 1-15 of 15
Compare
Product Number
Description
Pricing