Skip to Content
Merck
β-Glycerol phosphate disodium salt pentahydrate ≥98.0% (NT)

β-Glycerol phosphate disodium salt pentahydrate

β-Glycerol phosphate disodium salt pentahydrate ≥98.0% (NT)
Synonym(s):
β-GP, β-glycerolphosphate, Disodium β-glycerol phosphate pentahydrate, glycerol-2-phosphate
Empirical Formula (Hill Notation):
C3H7Na2O6P · 5H2O
CAS Number:
Molecular Weight:
306.11
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing