
Benzyltrimethylammonium dichloroiodate

Synonym(s):
BTMA-ICl2, Benzyltrimethylammonium dichloroiodide
Linear Formula:
C6H5CH2N(ICl2)(CH3)3
CAS Number:
Molecular Weight:
348.05
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1