
Atrial Natriuretic Peptide rat

Synonym(s):
α-Atriopeptin, rat, α-rANP, ANF (123-150), rat, ANF 1-28, rANP
Empirical Formula (Hill Notation):
C128H205N45O39S2
CAS Number:
Molecular Weight:
3062.41
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Chemical Composition
Quality Segment
Showing 1-1 of 1