Asunaprevir
Synonym(s):
Asunaprevir, BMS650032, 3-Methyl-N-{[(2-methyl-2-propanyl)oxy]carbonyl}-L-valyl-(4R)-4-[(7-chloro-4-methoxy-1-isoquinolinyl)oxy]-N-{(1R,2S)-1-[(cyclopropylsulfonyl)carbamoyl]-2-vinylcyclopropyl}-L-prolinamide, ASV, BMS 650032, BMS-650032
Empirical Formula (Hill Notation):
C35H46ClN5O9S
CAS Number:
Molecular Weight:
748.29
Products
Brand
Color
Feature
Formula Weight
Physical Form
Activity Spectrum
Quality Segment
Mode of Action
Showing 1-2 of 2
Compare
Product Number
Description
Pricing