![Ascorbic acid Impurity C Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/140/715/b9890ffe-5109-48f4-883a-5ea3d997673b/640/b9890ffe-5109-48f4-883a-5ea3d997673b.jpg)
Ascorbic acid impurity C
![Ascorbic acid Impurity C Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/140/715/b9890ffe-5109-48f4-883a-5ea3d997673b/640/b9890ffe-5109-48f4-883a-5ea3d997673b.jpg)
Synonym(s):
D-xylo-Hex-2-ulosonic acid
Empirical Formula (Hill Notation):
C6H10O7
CAS Number:
Molecular Weight:
194.14
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2