
Anti-TUBA4A (TUBA1) Antibody

Synonym(s):
Monoclonal Anti-α-Tubulin, Monoclonal Anti-α-Tubulin antibody produced in mouse, Anti-FLJ30169, Anti-H2-α, Anti-TUBA1, Anti-Tubulin, α 4a, FLJ30169, H2-ALPHA, TUBA1, Anti-H2ALPHA, Anti-tubulin, alpha 1 (testis specific)
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Isotype
Physical Form
Antibody Form
Conjugate
Quality Segment
Application
Post Translational Modification
Species Reactivity
Technique
Showing 1-10 of 10
Compare
Product Number
Description
Pricing