![Anti-SCHIP1 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/749/132/c448c54c-184e-47cb-8f23-d374d7958a27/640/c448c54c-184e-47cb-8f23-d374d7958a27.jpg)
Anti-SCHIP1 antibody produced in rabbit
![Anti-SCHIP1 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/749/132/c448c54c-184e-47cb-8f23-d374d7958a27/640/c448c54c-184e-47cb-8f23-d374d7958a27.jpg)
Synonym(s):
Anti-SCHIP-1, SCHIP-1
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing