Anti-Presenilin-2, N-terminal antibody produced in rabbit
Synonym(s):
Anti-AD3L, Anti-AD4, Anti-PS2, Anti-PSEN2, Anti-Presenilin 2 (Alzheimer 4), Anti-STM2
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing