Anti-polyHistidine−Peroxidase antibody, Mouse monoclonal
Synonym(s):
Monoclonal Anti-polyHistidine, Monoclonal 6 His epitope tag, Monoclonal 6xHis-tag, Monoclonal HHHHHH epitope tag, Monoclonal Hexa His tag, Monoclonal His-tag, Monoclonal His6 tag, Monoclonal Histidine tagged, Monoclonal Poly-His-tag
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Conjugate
Quality Segment
Package Size
Post Translational Modification
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing