Skip to Content
Merck
Anti-MUC16 antibody produced in rabbit affinity isolated antibody

Anti-MUC16 antibody produced in rabbit

Anti-MUC16 antibody produced in rabbit affinity isolated antibody
Synonym(s):
CA-125, Ovarian cancer-related tumor marker CA125, Ovarian carcinoma antigen CA125

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique