![Anti-ELF3 antibody produced in rabbit Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/649/916/e1e09340-b9f3-42c6-9e62-5c770f72e44b/640/e1e09340-b9f3-42c6-9e62-5c770f72e44b.jpg)
Anti-ELF3 antibody produced in rabbit
![Anti-ELF3 antibody produced in rabbit Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/649/916/e1e09340-b9f3-42c6-9e62-5c770f72e44b/640/e1e09340-b9f3-42c6-9e62-5c770f72e44b.jpg)
Synonym(s):
Anti-E74-like factor 3 antibody produced in rabbit, Anti-ets domain transcription factor, epithelial-specific antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1