Anti-EDNRA antibody produced in rabbit
Synonym(s):
ET-A, ETA-R, Endothelin A receptor, Endothelin-1 receptor, hET-AR
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing