![Anti-CTAGE5 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/330/159/6b4c2743-31f9-45b4-bd79-a17c598fd4a7/640/6b4c2743-31f9-45b4-bd79-a17c598fd4a7.jpg)
Anti-CTAGE5 antibody produced in rabbit
![Anti-CTAGE5 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/330/159/6b4c2743-31f9-45b4-bd79-a17c598fd4a7/640/6b4c2743-31f9-45b4-bd79-a17c598fd4a7.jpg)
Synonym(s):
Anti-Cutaneous T-cell lymphoma-associated antigen 5 antibody produced in rabbit, Anti-Meningioma-expressed antigen 6/11 antibody produced in rabbit, Anti-cTAGE family member 5 antibody produced in rabbit, Anti-cTAGE-5 protein antibody produced in rabbit, Anti-CTAGE family, member 5, Anti-MEA6, Anti-MGEA, Anti-MGEA11, Anti-MGEA6
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing