![Anti-β-Tubulin−Cy3 antibody, Mouse monoclonal clone TUB 2.1, purified from hybridoma cell culture](/deepweb/assets/sigmaaldrich/product/images/658/441/16ac04f9-344b-471f-9726-96ded247562d/640/16ac04f9-344b-471f-9726-96ded247562d.jpg)
Anti-β-Tubulin−Cy3 antibody, Mouse monoclonal
![Anti-β-Tubulin−Cy3 antibody, Mouse monoclonal clone TUB 2.1, purified from hybridoma cell culture](/deepweb/assets/sigmaaldrich/product/images/658/441/16ac04f9-344b-471f-9726-96ded247562d/640/16ac04f9-344b-471f-9726-96ded247562d.jpg)
Synonym(s):
Monoclonal Anti-β-Tubulin
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing