![Anti-BrdU antibody, Mouse monoclonal clone BU-33, purified from hybridoma cell culture](/deepweb/assets/sigmaaldrich/product/images/263/412/b38f0613-ac72-475d-bc73-c054f1e40cfa/640/b38f0613-ac72-475d-bc73-c054f1e40cfa.jpg)
Anti-BrdU antibody, Mouse monoclonal
![Anti-BrdU antibody, Mouse monoclonal clone BU-33, purified from hybridoma cell culture](/deepweb/assets/sigmaaldrich/product/images/263/412/b38f0613-ac72-475d-bc73-c054f1e40cfa/640/b38f0613-ac72-475d-bc73-c054f1e40cfa.jpg)
Synonym(s):
Monoclonal Anti-BrdU antibody produced in mouse, Anti-Bromodeoxyuridine
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Conjugate
Quality Segment
Package Size
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing