![Adenosine 3′-phosphate 5′-phosphosulfate lithium salt hydrate ≥60%](/deepweb/assets/sigmaaldrich/product/structures/232/387/a9e6ff0e-a388-47bd-9669-f13dbcbbce86/640/a9e6ff0e-a388-47bd-9669-f13dbcbbce86.png)
Adenosine 3′-phosphate 5′-phosphosulfate lithium salt hydrate
![Adenosine 3′-phosphate 5′-phosphosulfate lithium salt hydrate ≥60%](/deepweb/assets/sigmaaldrich/product/structures/232/387/a9e6ff0e-a388-47bd-9669-f13dbcbbce86/640/a9e6ff0e-a388-47bd-9669-f13dbcbbce86.png)
Synonym(s):
3′-Phosphoadenosine 5′-phosphosulfate lithium salt, APPS, Active sulfate, PAPS
Empirical Formula (Hill Notation):
C10H15N5O13P2S · xLi+ · yH2O
CAS Number:
Molecular Weight:
507.26 (free acid basis)
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing