![α-Chaconine phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/251/279/9d234b7e-d7c9-4924-b47c-e80e395f93ab/640/9d234b7e-d7c9-4924-b47c-e80e395f93ab.png)
α-Chaconine
![α-Chaconine phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/251/279/9d234b7e-d7c9-4924-b47c-e80e395f93ab/640/9d234b7e-d7c9-4924-b47c-e80e395f93ab.png)
Empirical Formula (Hill Notation):
C45H73NO14
CAS Number:
Molecular Weight:
852.06
Products
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing