![6-Ethoxy-2-hydroxyquinoline-3-carbaldehyde AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/159/497/4a2d4c45-b574-4329-a02c-5f078115112c/640/4a2d4c45-b574-4329-a02c-5f078115112c.png)
6-Ethoxy-2-hydroxyquinoline-3-carbaldehyde
![6-Ethoxy-2-hydroxyquinoline-3-carbaldehyde AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/159/497/4a2d4c45-b574-4329-a02c-5f078115112c/640/4a2d4c45-b574-4329-a02c-5f078115112c.png)
Empirical Formula (Hill Notation):
C12H11NO3
CAS Number:
Molecular Weight:
217.22
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing