![6β-Hydroxytropinone phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/148/192/aaad5b0c-d781-4022-b638-7674bf32672a/640/aaad5b0c-d781-4022-b638-7674bf32672a.png)
6β-Hydroxytropinone
![6β-Hydroxytropinone phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/148/192/aaad5b0c-d781-4022-b638-7674bf32672a/640/aaad5b0c-d781-4022-b638-7674bf32672a.png)
Synonym(s):
exo-6-Hydroxytropinone
Empirical Formula (Hill Notation):
C8H13NO2
CAS Number:
Molecular Weight:
155.19
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing