
(6,7-Dimethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-methanol

Empirical Formula (Hill Notation):
C12H17NO3
Molecular Weight:
223.27
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing