Skip to Content
Merck
5-Maleimido-eosin for fluorescence, ≥93% (HPLC)

5-Maleimido-eosin

5-Maleimido-eosin for fluorescence, ≥93% (HPLC)
Synonym(s):
Eosin-5-maleimide
Empirical Formula (Hill Notation):
C24H9Br4NO7
CAS Number:
Molecular Weight:
742.95

Products

Product Category
Brand
Formula Weight
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing