Skip to Content
Merck
4-tert-Butylcalix[6]arene 95%

4-tert-Butylcalix[6]arene

4-tert-Butylcalix[6]arene 95%
Empirical Formula (Hill Notation):
C66H84O6
CAS Number:
Molecular Weight:
973.37

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing