Skip to Content
Merck
4-Nitrophenyl butyrate ≥98%

4-Nitrophenyl butyrate

4-Nitrophenyl butyrate ≥98%
Synonym(s):
p-Nitrophenyl butyrate, Butyric acid 4-nitrophenyl ester
Empirical Formula (Hill Notation):
C10H11NO4
CAS Number:
Molecular Weight:
209.20

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1