![4-Chloromandelic acid AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/120/948/df5a51c0-8f92-4010-b0f7-16d71f3ee8bf/640/df5a51c0-8f92-4010-b0f7-16d71f3ee8bf.png)
4-Chloromandelic acid
![4-Chloromandelic acid AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/120/948/df5a51c0-8f92-4010-b0f7-16d71f3ee8bf/640/df5a51c0-8f92-4010-b0f7-16d71f3ee8bf.png)
Synonym(s):
4-Chloro-DL-mandelic acid, 4-Chloro-α-hydroxyphenylacetic acid
Empirical Formula (Hill Notation):
C8H7ClO3
CAS Number:
Molecular Weight:
186.59
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing