![4-Bromobenzocyclobutene 96%](/deepweb/assets/sigmaaldrich/product/structures/405/581/62c1f6dd-781a-47ae-88fc-bd07b5219b84/640/62c1f6dd-781a-47ae-88fc-bd07b5219b84.png)
4-Bromobenzocyclobutene
![4-Bromobenzocyclobutene 96%](/deepweb/assets/sigmaaldrich/product/structures/405/581/62c1f6dd-781a-47ae-88fc-bd07b5219b84/640/62c1f6dd-781a-47ae-88fc-bd07b5219b84.png)
Synonym(s):
3-Bromobicyclo[4.2.0]octa-1,3,5-triene, 4-Bromo-1,2-dihydrobenzocyclobutene
Empirical Formula (Hill Notation):
C8H7Br
CAS Number:
Molecular Weight:
183.05
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing