Skip to Content
Merck
4-(4-Methoxyphenyl)-2-butanone natural, ≥96%, FG

4-(4-Methoxyphenyl)-2-butanone

4-(4-Methoxyphenyl)-2-butanone natural, ≥96%, FG
Synonym(s):
Anisylacetone
Linear Formula:
CH3OC6H4CH2CH2COCH3
CAS Number:
Molecular Weight:
178.23

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-3 of 3