
4,4′-Dianilino-1,1′-binaphthyl-5,5′-disulfonic acid dipotassium salt

Synonym(s):
Bis-ANS
Empirical Formula (Hill Notation):
C32H22K2N2O6S2
CAS Number:
Molecular Weight:
672.85
Products
Product Category
Brand
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing