![3-Methoxythiophene 98%](/deepweb/assets/sigmaaldrich/product/structures/103/182/56340eb7-d521-4c10-9998-0047eb21433b/640/56340eb7-d521-4c10-9998-0047eb21433b.png)
3-Methoxythiophene
![3-Methoxythiophene 98%](/deepweb/assets/sigmaaldrich/product/structures/103/182/56340eb7-d521-4c10-9998-0047eb21433b/640/56340eb7-d521-4c10-9998-0047eb21433b.png)
Empirical Formula (Hill Notation):
C5H6OS
CAS Number:
Molecular Weight:
114.17
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing