Skip to Content
Merck
3,5-Di-tert-butyl-4-hydroxybenzyl alcohol certified reference material, pharmaceutical secondary standard

3,5-Di-tert-butyl-4-hydroxybenzyl alcohol

3,5-Di-tert-butyl-4-hydroxybenzyl alcohol certified reference material, pharmaceutical secondary standard
Linear Formula:
HOC6H2[C(CH3)3]2CH2OH
CAS Number:
Molecular Weight:
236.35

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade