Skip to Content
Merck
3-(4-Methylbenzylidene)camphor certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

3-(4-Methylbenzylidene)camphor

3-(4-Methylbenzylidene)camphor certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
1,7,7-Trimethyl-3-(4-methylbenzylidene)-norbornan-2-one, 1,7,7-Trimethyl-3-[(4-methylphenyl)methylene]-bicyclo[2.2.1]heptan-2-one, 4-MBC, 4-Methylbenzylidene camphor
Empirical Formula (Hill Notation):
C18H22O
CAS Number:
Molecular Weight:
254.37
EC Number:

Products

Brand
Formula Weight
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique