Skip to Content
Merck
2-(Trimethylsiloxy)-1,3-cyclohexadiene 95%

2-(Trimethylsiloxy)-1,3-cyclohexadiene

2-(Trimethylsiloxy)-1,3-cyclohexadiene 95%
Empirical Formula (Hill Notation):
C9H16OSi
CAS Number:
Molecular Weight:
168.31

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing