Skip to Content
Merck
2-Methoxyethyl acetate reagent grade, 98%

2-Methoxyethyl acetate

2-Methoxyethyl acetate reagent grade, 98%
Synonym(s):
1-Acetoxy-2-methoxyethane, Ethylene glycol monomethyl ether acetate, Methyl Cellosolve® acetate, 2-Methoxyethyl acetate
Linear Formula:
CH3COOCH2CH2OCH3
CAS Number:
Molecular Weight:
118.13

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Application
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing