2-Ethylhexyl trans-4-methoxycinnamate
Linear Formula:
CH3OC6H4CH=CHCO2CH2CH(C2H5)(CH2)3CH3
CAS Number:
Molecular Weight:
290.40
Products
Brand
Boiling Point (°C)
Formula Weight
Format
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing