2-Chlorotrityl hydrazine, polymer-bound
Synonym(s):
(α-Hydrazino-α-phenyl-2-chlorobenzyl)polystyrene, Hydrazine bound to 2-chloro-trityl resin
Products
Brand
Physical Form
Functional Group
Application
Cleavage Condition
Linker
Chain Type
Reaction Type
Showing 1-2 of 2