2-Bromoallyltrimethylsilane
Synonym(s):
2-Bromo-3-(trimethylsilyl)propene
Linear Formula:
H2C=C(Br)CH2Si(CH3)3
CAS Number:
Molecular Weight:
193.16
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing