Skip to Content
Merck
2-(4-Biphenylyl)-6-phenylbenzoxazole BioReagent, suitable for scintillation, ≥98.0% (TLC)

2-(4-Biphenylyl)-6-phenylbenzoxazole

2-(4-Biphenylyl)-6-phenylbenzoxazole BioReagent, suitable for scintillation, ≥98.0% (TLC)
Synonym(s):
PBBO
Empirical Formula (Hill Notation):
C25H17NO
CAS Number:
Molecular Weight:
347.41
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Special Grade
Showing 1-1 of 1