
2′,3′-Dideoxycytidine

Synonym(s):
ddC
Empirical Formula (Hill Notation):
C9H13N3O3
CAS Number:
Molecular Weight:
211.22
Products
Brand
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1