Skip to Content
Merck
2,2,3,4,4,4-Hexafluorobutyl methacrylate 98%

2,2,3,4,4,4-Hexafluorobutyl methacrylate

2,2,3,4,4,4-Hexafluorobutyl methacrylate 98%
Synonym(s):
HFBMA
Linear Formula:
CH2=C(CH3)COOCH2CF2CFHCF3
CAS Number:
Molecular Weight:
250.14

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing