(1R,2S)-cis-2-Aminocyclopentanol hydrochloride
Empirical Formula (Hill Notation):
C5H11NO · HCl
CAS Number:
Molecular Weight:
137.61
Products
Product Category
Brand
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing