1-Phenyl-1H-pyrazole-4-carboxaldehyde
Empirical Formula (Hill Notation):
C10H8N2O
CAS Number:
Molecular Weight:
172.18
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing