Skip to Content
Merck
1-Octanesulfonic acid sodium salt free-flowing, Redi-Dri™, ≥98%

1-Octanesulfonic acid sodium salt

1-Octanesulfonic acid sodium salt free-flowing, Redi-Dri™, ≥98%
Linear Formula:
CH3(CH2)7SO3Na
CAS Number:
Molecular Weight:
216.27

Products

Shipping
Product Category
Brand
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing