Skip to Content
Merck
1-Methyl-2-pyrrolidinone suitable for HPLC, ≥99%

1-Methyl-2-pyrrolidinone

1-Methyl-2-pyrrolidinone suitable for HPLC, ≥99%
Synonym(s):
1-Methyl-2-pyrrolidone, N-Methyl-2-pyrrolidone, NMP
Empirical Formula (Hill Notation):
C5H9NO
CAS Number:
Molecular Weight:
99.13
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing