![1-Butyl-3-methylimidazolium methyl sulfate ≥95%](/deepweb/assets/sigmaaldrich/product/structures/309/604/6a9da98c-9011-4815-bd1d-24bc3ac3bd46/640/6a9da98c-9011-4815-bd1d-24bc3ac3bd46.png)
1-Butyl-3-methylimidazolium methyl sulfate
![1-Butyl-3-methylimidazolium methyl sulfate ≥95%](/deepweb/assets/sigmaaldrich/product/structures/309/604/6a9da98c-9011-4815-bd1d-24bc3ac3bd46/640/6a9da98c-9011-4815-bd1d-24bc3ac3bd46.png)
Synonym(s):
BMIMMeSO4
Empirical Formula (Hill Notation):
C9H18N2O4S
CAS Number:
Molecular Weight:
250.32
Products
Brand
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing