16-Hexadecanolide
Synonym(s):
Juniperic acid lactone, NSC 33546
Empirical Formula (Hill Notation):
C16H30O2
CAS Number:
Molecular Weight:
254.41
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing