Skip to Content
Merck
1-[(2RS)-2-(2,4-Dichlorophenyl)-2-[[(2,5-dichlorophenyl)methyl]oxy]ethyl]-1H-imidazolium nitrate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

1-[(2RS)-2-(2,4-Dichlorophenyl)-2-[[(2,5-dichlorophenyl)methyl]oxy]ethyl]-1H-imidazolium nitrate

1-[(2RS)-2-(2,4-Dichlorophenyl)-2-[[(2,5-dichlorophenyl)methyl]oxy]ethyl]-1H-imidazolium nitrate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Miconazole Nitrate impurity G (PhEur) as Nitrate
Empirical Formula (Hill Notation):
C18H14Cl4N2O · HNO3
CAS Number:
Molecular Weight:
479.14

Products

Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade