1,2-Dichlorobenzene-d4 solution
Empirical Formula (Hill Notation):
C6D4Cl2
CAS Number:
Molecular Weight:
151.03
Products
Brand
Formula Weight
Product Line
Agency
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing