
10-O-Deacetylpaclitaxel

Synonym(s):
10-Deacetyltaxol
Empirical Formula (Hill Notation):
C45H49NO13
CAS Number:
Molecular Weight:
811.87
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing